Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130831.1 | NP_001124303.1 | 7 | 669 | ||
NM_003644.2 | NP_003635.2 | 3 | 418 | |||
NM_201432.1 | NP_958836.1 | 7 | 826 | |||
NM_201433.1 | NP_958839.1 | 7 | 856 | |||
XM_005256831.3 | XP_005256888.1 | 8 | 767 | |||
XM_005256832.3 | XP_005256889.1 | 7 | 637 | |||
XM_005256833.3 | XP_005256890.1 | 6 | 744 | |||
XM_005256834.3 | XP_005256891.1 | 6 | 570 | |||
XM_005256835.3 | XP_005256892.1 | 6 | 514 | |||
XM_011524044.2 | XP_011522346.1 | 7 | 661 | |||
XM_017025239.1 | XP_016880728.1 | 7 | 690 | |||
XM_017025240.1 | XP_016880729.1 | 6 | 473 | |||
XM_017025241.1 | XP_016880730.1 | 6 | 449 | |||
XM_017025242.1 | XP_016880731.1 | 6 | 623 | |||
GenBank mRNA | AB007854.1 | - | 7 | 624 | ||
AB209030.1 | - | 6 | 513 | |||
AJ224876.1 | - | 3 | 441 | |||
AK057761.1 | - | 4 | 488 | |||
AK225165.1 | - | 7 | 648 | |||
AK292987.1 | - | 7 | 642 | |||
AK293755.1 | - | 3 | 249 | |||
AK293834.1 | - | 3 | 430 | |||
AK294829.1 | - | 6 | 617 | |||
AK307987.1 | - | 8 | 678 | |||
AK309722.1 | - | 7 | 677 | |||
AK315162.1 | - | 3 | 431 | |||
AY327406.1 | - | 7 | 856 | |||
BC001152.1 | - | 7 | 649 | 3353809 | ||
BC006454.1 | - | 7 | 826 | 3547000 | ||
BT006891.1 | - | 6 | 503 |