Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003688.1 | NP_001003688.1 | 2 | 314 | ||
NM_005900.2 | NP_005891.1 | 2 | 490 | |||
XM_005262992.3 | XP_005263049.1 | 2 | 283 | |||
XM_006714217.2 | XP_006714280.1 | 2 | 345 | |||
XM_011531961.1 | XP_011530263.1 | 2 | 305 | |||
XM_011531962.1 | XP_011530264.1 | 2 | 350 | |||
XM_011531963.1 | XP_011530265.1 | 2 | 472 | |||
XM_011531964.1 | XP_011530266.1 | 2 | 502 | |||
GenBank mRNA | AK293055.1 | - | 2 | 616 | ||
BC001878.1 | - | 2 | 415 | 3546771 | ||
BQ962660.1 | - | 2 | 382 | |||
BT007386.1 | - | 1 | 74 | |||
BU163870.1 | - | 2 | 501 | |||
U54826.1 | - | 2 | 314 | |||
U57456.1 | - | 1 | 226 | |||
U59423.1 | - | 2 | 506 | |||
U59912.1 | - | 2 | 315 |
Gene Symbol: SMAD1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW087665.1 | - | 1 | 75 |