Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031553.2 | NP_113741.1 | 4 | 329 | ||
XM_006241196.2 | XP_006241258.1 | 4 | 347 | |||
GenBank mRNA | BC089791.1 | - | 4 | 329 | 7380395 | |
M55045.1 | - | 3 | 198 |