Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031565.2 | NP_113753.2 | 4 | 636 | ||
GenBank mRNA | EX493673.1 | - | 4 | 428 | ||
X81395.1 | - | 4 | 501 |