Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006186.3 | NP_006177.1 | 3 | 703 | ||
XM_005246621.3 | XP_005246678.1 | 2 | 2487 | |||
XM_005246622.3 | XP_005246679.1 | 3 | 471 | |||
XM_006712553.3 | XP_006712616.1 | 2 | 2487 | |||
XM_011511246.1 | XP_011509548.1 | 2 | 2487 | |||
XM_017004219.1 | XP_016859708.1 | 2 | 2046 | |||
XM_017004220.1 | XP_016859709.1 | 3 | 2611 | |||
GenBank mRNA | AK223625.1 | - | 3 | 644 | ||
AK291456.1 | - | 3 | 464 | |||
AK299439.1 | - | 2 | 517 | |||
BC009288.2 | - | 3 | 633 | 4298967 | ||
BC066890.1 | - | 3 | 434 | 4828460 | ||
BM922519.1 | - | 3 | 628 | |||
HQ692824.1 | - | 1 | 281 | |||
HQ709176.1 | - | 1 | 281 | |||
KU178238.1 | - | 1 | 281 | |||
KU178239.1 | - | 1 | 281 | |||
S77154.1 | - | 3 | 290 | |||
X75918.1 | - | 3 | 598 |