Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031332.1 | NP_112622.1 | 7 | 1058 | ||
XM_008760243.1 | XP_008758465.1 | 6 | 873 | |||
GenBank mRNA | AB017446.1 | - | 7 | 1058 | ||
BC081777.1 | - | 7 | 1056 | 7109603 | ||
FQ209755.1 | - | 1070 |