Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031337.2 | NP_112627.2 | 6 | 1180 | ||
XM_006236676.1 | XP_006236738.1 | 5 | 841 | |||
XM_006236677.1 | XP_006236739.1 | 6 | 983 | |||
XM_006236679.2 | XP_006236741.1 | 5 | 974 | |||
XM_006236680.1 | XP_006236742.1 | 5 | 841 | |||
XM_006236682.1 | XP_006236744.1 | 4 | 668 | |||
XM_006236684.2 | XP_006236746.1 | 4 | 862 | |||
XM_008762996.1 | XP_008761218.1 | 5 | 1003 | |||
XM_008762997.1 | XP_008761219.1 | 4 | 891 | |||
GenBank mRNA | AB018049.1 | - | 5 | 846 | ||
BC078798.1 | - | 6 | 1180 | 7106217 |