Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006289.3 | NP_006280.3 | 3 | 536 | ||
GenBank mRNA | AB028950.2 | - | 3 | 511 | ||
AF078828.1 | - | 3 | 308 | |||
AF177198.1 | - | 3 | 272 | |||
BC042923.1 | - | 3 | 322 | 5576203 | ||
Z50170.1 | - | 2 | 172 |