Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031510.1 | NP_113698.1 | 4 | 592 | ||
XM_006245049.2 | XP_006245111.1 | 5 | 650 | |||
XM_008767076.1 | XP_008765298.1 | 5 | 682 | |||
GenBank mRNA | DQ403076.1 | - | 4 | 482 | ||
FQ214242.1 | - | 5 | 673 | |||
L35317.1 | - | 4 | 592 |