Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031542.2 | NP_113730.2 | 10 | 1939 | ||
GenBank mRNA | U89653.1 | - | 10 | 1938 |