Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phkg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031573.1 | NP_113761.1 | 5, 6 | 459 | ||
GenBank mRNA | X07320.1 | - | 5, 6 | 459 |