Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031583.2 | NP_113771.2 | 13 | 1240 | ||
GenBank mRNA | FQ225069.1 | - | 12 | 1077 | ||
FQ231827.1 | - | 12 | 1077 | |||
FQ231919.1 | - | 13 | 1253 | |||
FQ233317.1 | - | 13 | 1255 | |||
FQ234437.1 | - | 13 | 1253 | |||
U82626.1 | - | 13 | 1207 |