Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc37a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031589.2 | NP_113777.2 | 6, 7 | 1159 | ||
XM_008766151.1 | XP_008764373.1 | 5, 6 | 920 | |||
XM_008766152.1 | XP_008764374.1 | 5, 6 | 985 | |||
XM_008766153.1 | XP_008764375.1 | 5, 6 | 920 | |||
GenBank mRNA | AF080468.1 | - | 5, 6 | 989 | ||
BC070889.1 | - | 6, 7 | 1159 | 7098680 |