Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sart1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031596.1 | NP_113784.1 | 2, 3 | 429 | ||
GenBank mRNA | AB014722.1 | - | 2, 3 | 429 | ||
BC083551.1 | - | 2, 3 | 402 | 7134496 |