Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctla4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031674.1 | NP_113862.1 | 2 | 398 | ||
XM_006245036.2 | XP_006245098.1 | 2 | 516 | |||
GenBank mRNA | U37121.1 | - | 2 | 398 | ||
U90271.1 | - | 2 | 398 |