Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031697.1 | NP_113885.1 | 6, 7 | 478 | ||
XM_006238715.2 | XP_006238777.1 | 6, 7 | 915 | |||
XM_006238716.2 | XP_006238778.1 | 5, 6 | 881 | |||
XM_008764066.1 | XP_008762288.1 | 6, 7 | 3405 | |||
XM_008764067.1 | XP_008762289.1 | 6, 7 | 498 | |||
GenBank mRNA | M97754.1 | - | 6, 7 | 478 |