Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031594.1 | NP_113782.1 | 2, 3 | 390 | ||
GenBank mRNA | BC078792.1 | - | 2, 3 | 331 | 7123247 | |
U32497.1 | - | 2, 3 | 367 | |||
U47031.1 | - | 2, 3 | 320 | |||
X87763.1 | - | 2, 3 | 390 | |||
X91200.1 | - | 2, 3 | 305 | |||
X93565.1 | - | 2, 3 | 477 |