Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031660.1 | NP_113848.1 | 3 | 266 | ||
XM_006243410.2 | XP_006243472.1 | 4 | 377 | |||
GenBank mRNA | AJ005982.1 | - | 3 | 266 | ||
BC058461.1 | - | 4 | 312 | 6919944 | ||
FQ213926.1 | - | 2 | 347 | |||
FQ221182.1 | - | 4 | 316 | |||
FQ222843.1 | - | 4 | 315 |
Gene Symbol: LOC100360828 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729972.4 | XP_002730018.2 | 1 | 303 |