Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc28a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031664.1 | NP_113852.1 | 3 | 396 | ||
GenBank mRNA | AY029302.1 | - | 3 | 377 | ||
EU032627.2 | - | 3 | 464 | |||
U25055.1 | - | 3 | 396 | |||
U66723.1 | - | 3 | 367 |
Gene Symbol: LOC102554663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762219.1 | XP_008760441.1 | 2 | 921 | ||
XM_008762220.1 | XP_008760442.1 | 3 | 365 | |||
XM_008762221.1 | XP_008760443.1 | 3 | 362 |