Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sncg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031688.1 | NP_113876.1 | 3 | 307 | ||
GenBank mRNA | AY518351.1 | - | 3 | 257 | ||
X86789.1 | - | 3 | 307 |