Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031698.1 | NP_113886.1 | 3, 4 | 312 | ||
XM_006235379.1 | XP_006235441.1 | 3, 4 | 344 | |||
GenBank mRNA | BC060556.1 | - | 4, 5 | 322 | 6919638 | |
X55298.1 | - | 3, 4 | 312 |