Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004706.3 | NP_004697.2 | 3 | 204 | ||
NM_198977.1 | NP_945328.1 | 3 | 204 | |||
NM_199002.1 | NP_945353.1 | 3 | 229 | |||
XM_005259386.4 | XP_005259443.1 | 3 | 556 | |||
XM_005259390.4 | XP_005259447.1 | 3 | 552 | |||
XM_011527468.2 | XP_011525770.1 | 3 | 552 | |||
GenBank mRNA | BC011726.2 | - | 3 | 179 | 4138058 | |
BC034013.1 | - | 3 | 160 | 3451036 | ||
BC067262.1 | - | 3 | 147 | 6502165 | ||
BM147791.1 | - | 3 | 205 | |||
BQ944132.1 | - | 3 | 317 | |||
U64105.1 | - | 3 | 149 |