Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031623.1 | NP_113811.1 | 2, 3 | 395 | ||
GenBank mRNA | AF076619.1 | - | 2, 3 | 395 |