Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031639.1 | NP_113827.1 | 16 | 2276 | ||
XM_006257080.2 | XP_006257142.1 | 16 | 2603 | |||
XM_006257081.2 | XP_006257143.1 | 15 | 2548 | |||
XM_006257082.2 | XP_006257144.1 | 15 | 2511 | |||
XM_006257083.2 | XP_006257145.1 | 14 | 2455 | |||
XM_006257084.2 | XP_006257146.1 | 9 | 1003 | |||
XM_006257085.2 | XP_006257147.1 | 7 | 1394 | |||
XM_008773267.1 | XP_008771489.1 | 15 | 2563 | |||
XM_008773268.1 | XP_008771490.1 | 9 | 912 | |||
GenBank mRNA | U50147.1 | - | 16 | 2276 |