Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004739.3 | NP_004730.2 | 3 | 515 | ||
XM_017018561.1 | XP_016874050.1 | 3 | 367 | |||
GenBank mRNA | AB016591.1 | - | 3 | 448 | ||
AF295807.1 | - | 3 | 125 | |||
AK301569.1 | - | 3 | 350 | |||
AL561056.3 | - | 3 | 514 | |||
BC023656.2 | - | 3 | 311 | |||
BC053650.1 | - | 3 | 333 | 5531965 |