Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031656.2 | NP_113844.1 | 3 | 186 | ||
XM_008767863.1 | XP_008766085.1 | 4 | 410 | |||
XM_008767864.1 | XP_008766086.1 | 4 | 410 | |||
XM_008767865.1 | XP_008766087.1 | 4 | 410 | |||
GenBank mRNA | AF033109.1 | - | 3 | 191 |