Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031665.2 | NP_113853.1 | 4 | 487 | ||
GenBank mRNA | BC081769.1 | - | 4 | 487 | 7134641 | |
U56815.1 | - | 4 | 463 |