Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031682.1 | NP_113870.1 | 2 | 186 | ||
GenBank mRNA | AF049878.1 | - | 2 | 174 | ||
AF069770.1 | - | 2 | 186 | |||
BC158587.1 | - | 2 | 180 | 9089087 |