Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318204.1 | NP_001305133.1 | 1, 2 | 304 | ||
NM_004560.3 | NP_004551.2 | 1, 2 | 304 | |||
GenBank mRNA | BC051273.1 | - | 1, 2 | 116 | ||
BC130522.1 | - | 1, 2 | 192 | 40146553 | ||
M97639.1 | - | 1, 2 | 304 |