Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031723.3 | NP_113911.2 | 3 | 352 | ||
XM_008759558.1 | XP_008757780.1 | 3 | 308 | |||
XM_008759559.1 | XP_008757781.1 | 3 | 572 | |||
GenBank mRNA | BC060554.1 | - | 3 | 356 | 6921802 | |
FQ220501.1 | - | 3 | 356 | |||
FQ221649.1 | - | 1 | 59 | |||
L11319.1 | - | 3 | 339 |