Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031769.1 | NP_113957.1 | 1, 2 | 95 | ||
GenBank mRNA | AF020346.1 | - | 1, 2 | 95 | ||
FQ213832.1 | - | 1, 2 | 199 |
Gene Symbol: RGD1566085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772854.1 | XP_008771076.1 | 1, 2 | 225 |