Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031793.1 | NP_113981.1 | 6 | 496 | ||
XM_006234324.2 | XP_006234386.1 | 6 | 872 | |||
XM_006234325.2 | XP_006234387.1 | 6 | 693 | |||
XM_006234326.2 | XP_006234388.1 | 6 | 816 | |||
XM_006234327.2 | XP_006234389.1 | 5 | 768 | |||
XM_006234328.2 | XP_006234390.1 | 6 | 763 | |||
XM_006234330.2 | XP_006234392.1 | 5 | 589 | |||
XM_006234331.2 | XP_006234393.1 | 5 | 715 | |||
XM_006234333.2 | XP_006234395.1 | 5 | 729 | |||
XM_006234334.2 | XP_006234396.1 | 5 | 550 | |||
XM_006234335.2 | XP_006234397.1 | 5 | 676 | |||
XM_008761930.1 | XP_008760152.1 | 5 | 792 | |||
XM_008761931.1 | XP_008760153.1 | 5 | 753 | |||
GenBank mRNA | AF043642.1 | - | 6 | 496 | ||
BC085723.1 | - | 5 | 542 | 7123868 | ||
FQ225838.1 | - | 11 | ||||
FQ230740.1 | - | 11 | ||||
FQ230786.1 | - | 11 | ||||
FQ232477.1 | - | 11 |