Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031715.1 | NP_113903.1 | 645 | |||
GenBank mRNA | BC094212.1 | - | 6 | 632 | 7314667 | |
D21869.1 | - | 6 | 589 | |||
FN433010.1 | - | 6 | 610 | |||
FQ211719.1 | - | 771 | ||||
U25651.1 | - | 645 |