Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031730.2 | NP_113918.2 | 1 | 660 | ||
GenBank mRNA | M59980.1 | - | 1 | 339 | ||
S64320.1 | - | 1 | 658 |