Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031742.1 | NP_113930.1 | 3 | 463 | ||
XM_006250491.2 | XP_006250553.1 | 3 | 480 | |||
GenBank mRNA | Z34264.1 | - | 3 | 463 |