Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031793.1 | NP_113981.1 | 6 | 555 | ||
XM_006234324.2 | XP_006234386.1 | 6 | 931 | |||
XM_006234325.2 | XP_006234387.1 | 6 | 752 | |||
XM_006234326.2 | XP_006234388.1 | 6 | 875 | |||
XM_006234327.2 | XP_006234389.1 | 5 | 827 | |||
XM_006234328.2 | XP_006234390.1 | 6 | 822 | |||
XM_006234330.2 | XP_006234392.1 | 5 | 648 | |||
XM_006234331.2 | XP_006234393.1 | 5 | 774 | |||
XM_006234333.2 | XP_006234395.1 | 5 | 788 | |||
XM_006234334.2 | XP_006234396.1 | 5 | 609 | |||
XM_006234335.2 | XP_006234397.1 | 5 | 735 | |||
XM_008761930.1 | XP_008760152.1 | 5 | 851 | |||
XM_008761931.1 | XP_008760153.1 | 5 | 812 | |||
GenBank mRNA | AF043642.1 | - | 6 | 555 | ||
BC085723.1 | - | 5 | 601 | 7123868 | ||
FQ225838.1 | - | 1 | 70 | |||
FQ230740.1 | - | 1 | 70 | |||
FQ232477.1 | - | 1 | 70 |