Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clns1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031719.1 | NP_113907.1 | 5 | 562 | ||
XM_006229792.2 | XP_006229854.1 | 5 | 625 | |||
GenBank mRNA | BC060555.1 | - | 5 | 606 | 6918578 | |
D13985.1 | - | 5 | 632 | |||
D17698.1 | - | 5 | 545 | |||
FQ212666.1 | - | 3 | 244 | |||
FQ232880.1 | - | 3 | 244 | |||
L26450.1 | - | 5 | 562 |