Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031740.1 | NP_113928.1 | 3 | 1095 | ||
GenBank mRNA | AF048687.1 | - | 3 | 1095 |