Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031743.2 | NP_113931.1 | 1 | 551 | ||
GenBank mRNA | AF021923.1 | - | 1 | 551 |