Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031776.2 | NP_113964.2 | 5, 6 | 621 | ||
GenBank mRNA | AF026472.2 | - | 5, 6 | 678 | ||
AF245172.1 | - | 5, 6 | 621 | |||
FQ233257.1 | - | 5, 6 | 643 | |||
FQ233523.1 | - | 5, 6 | 649 |