Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031779.2 | NP_113967.1 | 7 | 1929 | ||
XM_006231185.2 | XP_006231247.1 | 5 | 2029 | |||
XM_008760312.1 | XP_008758534.1 | 6 | 2062 | |||
GenBank mRNA | AF029105.1 | - | 2149 |