Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031780.1 | NP_113968.1 | 7 | 1477 | ||
GenBank mRNA | AF029107.1 | - | 7 | 1477 |