Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABCD4 SIRNA Assays ›
Gene Symbol:
ABCD4
Gene Name:
ATP binding cassette subfamily D member 4
Gene Aliases:
ABC41, EST352188, MAHCJ, P70R, P79R, PMP69, PXMP1L
Chromosome Location:
Chr.14: 74285277 - 74303064 on Build GRCh38
UniGene ID:
Hs.94395
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
5441
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABCD4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005050.3
NP_005041.1
6, 7
817
NR_003256.2
5, 6
689
XM_005267939.3
XP_005267996.1
6, 7
745
XM_005267940.3
XP_005267997.1
5, 6
624
XM_005267941.4
XP_005267998.1
5, 6
579
XM_005267942.4
XP_005267999.1
4, 5
466
XM_005267946.3
XP_005268003.1
4, 5
461
XM_005267949.3
XP_005268006.1
5, 6
675
XM_006720223.2
XP_006720286.1
5, 6
629
XM_011537041.2
XP_011535343.1
4, 5
487
XM_017021531.1
XP_016877020.1
6, 7
750
XM_017021534.1
XP_016877023.1
4, 5
431
XM_017021535.1
XP_016877024.1
6, 7
750
XM_017021536.1
XP_016877025.1
5, 6
570
XM_017021537.1
XP_016877026.1
4, 5
453
XM_017021538.1
XP_016877027.1
5, 6
571
XM_017021539.1
XP_016877028.1
5, 6
554
XM_017021540.1
XP_016877029.1
4, 5
423
XM_017021543.1
XP_016877032.1
4, 5
431
XM_017021544.1
XP_016877033.1
6, 7
750
GenBank mRNA
AF009746.1
-
6, 7
716
AK291332.1
-
6, 7
687
AK308382.1
-
5, 6
570
BC012815.2
-
6, 7
671
2960427
BQ893869.1
-
3, 4
275
BT007412.1
-
6, 7
665
BX247984.1
-
4, 5
486
BX248053.1
-
5, 6
617
CR457104.1
-
6, 7
665
Back To Top
More Information
Panther Classification:
Molecular Function -
ATP-binding cassette (ABC) transporter
Gene Ontology Categories:
Function(s)
Process(es)
cobalamin metabolic process
transmembrane transport
ATP binding
ATPase activity, coupled to transmembrane movement of substances
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline