Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031978.1 | NP_114184.1 | 4 | 273 | ||
GenBank mRNA | AJ006340.1 | - | 4 | 273 | ||
BC087654.1 | - | 4 | 358 | |||
BC105883.1 | - | 4 | 319 | |||
BC128734.1 | - | 4 | 318 | |||
BC166420.1 | - | 4 | 314 |