Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6kb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031985.1 | NP_114191.1 | 1 | 95 | ||
XM_006247111.2 | XP_006247173.1 | 1 | 232 | |||
XM_008768120.1 | XP_008766342.1 | 1 | 496 | |||
GenBank mRNA | BC089789.1 | - | 1 | 122 | ||
M58340.1 | - | 1 | 95 |