Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032063.2 | NP_114452.1 | 2 | 387 | ||
GenBank mRNA | U78889.2 | - | 2 | 387 |