Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113371.1 | NP_001106842.1 | 149 | |||
NM_001113372.1 | NP_001106843.1 | 149 | ||||
NM_032071.2 | NP_114460.1 | 204 | ||||
GenBank mRNA | AY034050.1 | - | 149 | |||
AY034051.1 | - | 149 | ||||
U90312.1 | - | 204 |