Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032079.2 | NP_114468.2 | 1, 2 | 106 | ||
GenBank mRNA | BC087010.1 | - | 1, 2 | 106 | 7191358 | |
U95727.1 | - | 1, 2 | 140 |