Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031825.1 | NP_114013.1 | 1, 2 | 171 | ||
XM_006234935.2 | XP_006234997.1 | 2, 3 | 460 | |||
XM_008762202.1 | XP_008760424.1 | 1, 2 | 361 | |||
XM_008762203.1 | XP_008760425.1 | 2, 3 | 455 | |||
GenBank mRNA | AF135059.1 | - | 1, 2 | 171 |