Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270800.1 | NP_001257729.1 | 3, 4 | 333 | ||
NM_001270801.1 | NP_001257730.1 | 3, 4 | 342 | |||
NM_031830.2 | NP_114018.1 | 3, 4 | 333 | |||
XM_006246952.1 | XP_006247014.1 | 4, 5 | 954 | |||
XM_008768119.1 | XP_008766341.1 | 2, 3 | 320 | |||
GenBank mRNA | AF023302.1 | - | 3, 4 | 312 | ||
AF023303.1 | - | 3, 4 | 312 | |||
AF023304.1 | - | 2, 3 | 230 | |||
CB581049.1 | - | 3, 4 | 333 | |||
U64999.1 | - | 1 | 185 |